Learn More
TARGETMOL CHEMICALS INC LEM-14-1189 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T729795MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Lem-14-1189 a LEM-14 derivative is a potent NSD inhibitor of the nuclear receptor binding SET domain and has inhibitory effects on NSD1 NSD2 and NSD3 with IC50 of 418 uM 111 uM and 60 uM respectively. LEM-14-1189 has potential anticancer activity and can be used to study multiple myeloma (MM) and diseases of the blood system. purity: 97%
Catalog No. 50-257-9226