Learn More
Medchemexpress LLC LEM-14 50mg | 1814881-70-3 | 50 MG

Supplier: Medchemexpress LLC HY11434050MG
LEM-14 is a research-grade small-molecule inhibitor of NSD2 (histone methyltransferase) supplied for laboratory research use only and not intended for human use.
- Potent and selective NSD2 inhibitor.
- Reported IC50 132 μM against NSD2.
- Molecular weight 478.56 g/mol.
- Chemical formula C25H26N4O4S.
- Purity approximately 98.0%.
- Available as a 50 mg solid and as a 10 mM solution in DMSO.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.