Learn More
Medchemexpress LLC LEM-14 5mg | 1814881-70-3 | 5 MG

Supplier: Medchemexpress LLC HY1143405MG
LEM-14 is a small-molecule inhibitor of the histone methyltransferase NSD2 used for laboratory research (CAS 1814881-70-3). It has a reported NSD2 IC50 of 132 μM and is supplied as a powder for preclinical studies.
- Small-molecule NSD2 inhibitor with reported IC50 = 132 μM.
- Molecular formula C25H26N4O4S; molecular weight 478.56.
- Purity typically 98.0% as specified by supplier.
- Supplied as a powder; store frozen to preserve stability.
- Intended for research use only, not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.