Learn More
TARGETMOL CHEMICALS INC LEMINOPRAZOLE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T278125MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Leminoprazole is an orally available H+K(+)-ATPase inhibitor that protects gastric mucosal cells from various cellular damages. purity: 99%
Catalog No. 50-258-0571