Learn More
Medchemexpress LLC Lenalidomide | 191732-72-6 | 99.9% | 5 G
SDP
Supplier:  Medchemexpress LLC HYA00035G
Encompass_Preferred
Lenalidomide is an orally active immunomodulator derived from Thalidomide, functioning as a molecular glue. It primarily targets ubiquitin E3 ligase cereblon (CRBN) to induce selective degradation of key lymphoid transcription factors, demonstrating significant activity against various cancers.
  • Acts as a ligand for ubiquitin E3 ligase cereblon (CRBN)
  • Causes selective ubiquitination and degradation of IKZF1 and IKZF3
  • Inhibits the growth of mature B-cell lymphomas and multiple myeloma
  • Induces IL-2 release and stimulates T cell proliferation
  • Suppresses pro-inflammatory cytokines (TNF-α, IL-1, IL-6, IL-12)
  • Enhances anti-inflammatory cytokine IL-10 production
  • Displays antiproliferative activity against human cell lines
Catalog No. 50-004-32896