Learn More
Medchemexpress LLC HY-A0003 1g Medchemexpress, Lenalidomide CC-5013 CAS:191732-72-6 Purity:98%
SDP
Supplier:  Medchemexpress LLC HYA00031G
Encompass_Preferred
Medchemexpress,HY-A0003 1g Lenalidomide CC-5013 CAS:191732-72-6 Formula:C13H13N3O3 Lenalidomide is a potent inhibitor of TNF-α used as a immunomodulatory drug. It has also been shown to have anti-angiogenic properties. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-9075