Learn More
Medchemexpress LLC Lenalidomide-4-aminomethyl hydrochloride | 444289-05-8 | 90.88% | 309.75 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY138882A
Encompass_Preferred
Lenalidomide-4-aminomethyl hydrochloride is a cereblon (CRBN) ligand derived from Lenalidomide, designed to recruit the CRBN protein. It serves as a component that can be linked to a target protein ligand via a linker, thereby forming a PROTAC. This compound is intended for research use only.
  • Cereblon (CRBN) ligand: Functions as a Lenalidomide-based cereblon ligand, facilitating the recruitment of the CRBN protein.
  • PROTAC formation: Can be connected to a ligand for a protein by a linker to create a PROTAC (proteolysis-targeting chimera).
  • Protein degradation mechanism: PROTACs, formed with this compound, utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Research applications: Suitable for various research purposes, particularly in areas involving protein degradation and E3 ligase ligands.
Catalog No. 50-003-48588