Learn More
Medchemexpress LLC Lenalidomide-4-aminomethyl hydrochloride | 444289-05-8 | 90.9% | 309.75 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY138882A50MG
Encompass_Preferred
Lenalidomide-4-aminomethyl hydrochloride is a Lenalidomide-based cereblon (CRBN) ligand used in the recruitment of CRBN protein. It can be connected to a ligand for protein by a linker to form PROTAC.
  • Cereblon (CRBN) ligand
  • Used in the recruitment of CRBN protein
  • Can be connected to a ligand for protein by a linker to form PROTAC
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-52395