Learn More
Medchemexpress LLC Lenalidomide-4-aminomethyl hydrochloride | 444289-05-8 | MFCD32703223 | 90.9% | C14H16ClN3O3 | 10MG
SDP
Supplier:  Medchemexpress LLC HY138882A10MG
Encompass_Preferred
Lenalidomide-4-aminomethyl hydrochloride is a lenalidomide-derived cereblon (CRBN) ligand used to recruit the CRBN E3 ubiquitin ligase for targeted protein degradation. It is commonly employed as the E3-binding moiety in PROTAC construction and can be conjugated to target ligands via a synthetic linker; supplied as the hydrochloride salt to aid handling and solubility.
  • Lenalidomide-based cereblon (CRBN) ligand for targeted protein degradation.
  • Suitable for use as the E3-binding moiety in PROTAC design and synthesis.
  • Hydrochloride salt form facilitates conjugation and handling in synthetic workflows.
  • Reported purity 90.9%; verify certificate of analysis for critical applications.
  • Provided for research use under appropriate laboratory safety conditions.
Catalog No. 50-002-60684