Learn More
Medchemexpress LLC Lenalidomide-4-br-7-fluoro | 2438239-34-8 | 97.0% | 341.13 | C13H10BrFN2O3 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW460227500MG
Encompass_Preferred
Lenalidomide-4-Br-7-fluoro is a brominated and fluorinated derivative of lenalidomide intended for research use as an E3 ligase ligand in PROTAC applications. It is supplied as a powder and characterized by molecular formula C13H10BrFN2O3 (MW 341.13) and CAS 2438239-34-8. Storage recommendations and analytical data are provided by the manufacturer.
Catalog No. 50-002-84213