Learn More
Medchemexpress LLC Lenalidomide 4'-PEG1-amine dihydrochloride | 2624336-86-1 | ≥95.0% | 419.30 g/mol | C17H24Cl2N4O4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY16112810MG
Encompass_Preferred
Lenalidomide 4'-PEG1-amine dihydrochloride is a lenalidomide-derived cereblon (CRBN) ligand bearing a short PEG1 spacer and a terminal primary amine, supplied as a dihydrochloride salt. It is intended for use as an E3 ligase recruiting building block in PROTAC synthesis and other conjugation chemistry where CRBN engagement is required.
  • Dihydrochloride salt with terminal primary amine suitable for linker attachment.
  • Short PEG1 linker provides minimal spacer length for conjugation.
  • Designed as a cereblon (CRBN) recruiting ligand for PROTACs and degraders.
  • Typically supplied as a solid for convenient handling and storage.
  • High purity (≥95% by HPLC) suitable for research applications.
Catalog No. 50-002-88748