Learn More
Medchemexpress LLC Lenalidomide 4'-PEG2-amine dihydrochloride | 2624336-88-3 | 99.0% | 463.36 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY16112910MM/1ML
Encompass_Preferred
Lenalidomide 4'-PEG2-amine dihydrochloride is a Lenalidomide-based Cereblon ligand that plays a role in the recruitment of CRBN protein. It can be connected to a ligand for protein by a linker to form PROTAC.
  • Functions as a Lenalidomide-based Cereblon ligand.
  • Recruits CRBN protein.
  • Connects to ligands for protein via a linker to form PROTACs.
Catalog No. 50-003-95206