Learn More
Medchemexpress LLC Lenalidomide 4'-PEG2-amine dihydrochloride | 2624336-88-3 | 99.0% | 463.36 g/mol | C19H28Cl2N4O5 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY16112910MG
Encompass_Preferred
Lenalidomide 4'-PEG2-amine dihydrochloride is a lenalidomide-derived Cereblon (CRBN) ligand functionalized with a PEG2-amine linker, supplied as the dihydrochloride salt for use in targeted protein degradation research. It is intended for conjugation to target ligands to construct PROTACs and other E3 ligase-recruiting molecules.
  • Lenalidomide-derived CRBN ligand functionalized with a PEG2-amine linker.
  • Designed for conjugation to target-binding ligands to enable PROTAC synthesis.
  • Dihydrochloride salt for improved handling in research workflows.
  • Molecular weight 463.36 g/mol and formula C19H28Cl2N4O5.
  • High purity (98.96%), suitable for research applications.
  • Available in mg-scale packages for lab-scale synthesis and screening.
Catalog No. 50-002-88599