Learn More
Medchemexpress LLC Lenalidomide-5-Br | 1010100-26-1 | MFCD32263142 | 99.9% | 323.15 | C13H11BrN2O3 | 50 G

Supplier: Medchemexpress LLC HYW07295450G
Lenalidomide-5-Br is a lenalidomide-derived cereblon (CRBN) ligand used to recruit the CRBN E3 ligase; it is supplied as a solid and is intended for use as a building block in PROTAC synthesis and other targeted protein degradation workflows.
- Chemical formula C13H11BrN2O3.
- Molecular weight 323.15 g/mol.
- Cas number 1010100-26-1.
- Purity 99.87% (reported by manufacturer).
- Solubility DMSO 12.5 mg/mL; water <0.1 mg/mL (insoluble).
- Recommended storage powder -20°C (3 years) or 4°C (2 years); in solution -80°C (6 months) or -20°C (1 month).
- Typical applications building block for PROTACs and CRBN recruitment in targeted protein degradation studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.