Learn More
Medchemexpress LLC Lenalidomide-5-Br | 1010100-26-1 | MFCD32263142 | 99.9% | 323.15 g/mol | C13H11BrN2O3 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW072954100G
Encompass_Preferred
Lenalidomide-5-Br is a brominated analog of lenalidomide that acts as a cereblon (CRBN) E3 ligase ligand used to recruit CRBN in targeted protein degradation and PROTAC synthesis. It is supplied as a high-purity solid reagent for synthetic and chemical biology applications.
  • Brominated lenalidomide analog suitable for CRBN recruitment.
  • High purity solid for synthetic chemistry and PROTAC assembly.
  • Molecular formula C13H11BrN2O3, molecular weight 323.15 g/mol.
  • Available in laboratory-scale quantities, including 100 G packs.
  • Documents available: data sheet, safety data sheet, and certificate of analysis.
Catalog No. 50-002-10240