Learn More
Medchemexpress LLC Lenalidomide-5-br | 1010100-26-1 | MFCD32263142 | 99.9% | 323.15 g/mol | C13H11BrN2O3 | 25 G

Supplier: Medchemexpress LLC 25GHYW072954
Lenalidomide-5-br is a brominated derivative of lenalidomide that acts as a cereblon (CRBN) ligand and is used to recruit CRBN in targeted protein degradation workflows such as PROTAC development. The compound is supplied as a solid with high stated purity and is available in multiple laboratory-scale package sizes.
- CRBN ligand suitable for PROTAC development and targeted protein degradation studies.
- Supplied as a solid with high stated purity for research use.
- Available in multiple package sizes for laboratory needs.
- Molecular formula C13H11BrN2O3 and molecular weight 323.15 g/mol.
- CAS number 1010100-26-1 for unambiguous identification.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.