Learn More
Medchemexpress LLC Lenalidomide 5'-piperazine hydrochloride | 2222120-31-0 | 99.9% | 364.83 | C17H21ClN4O3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW25993250MG
Encompass_Preferred
Lenalidomide 5'-piperazine hydrochloride is a research-grade cereblon (CRBN) ligand used in chemical biology and degrader (PROTAC) development. It provides a piperazine-functionalized handle for linker attachment and is supplied as a hydrochloride salt for research use only.
Catalog No. 50-002-81669