Learn More
Medchemexpress LLC Lenalidomide 5'-piperazine hydrochloride | 2222120-31-0 | MFCD32705051 | 99.9% | 364.83 g/mol | C17H21ClN4O3 | 100 MG

Supplier: Medchemexpress LLC HYW259932100MG
Lenalidomide 5'-piperazine hydrochloride is a lenalidomide-derived small molecule used as a cereblon (CRBN) ligand in targeted protein degradation and PROTAC research. It is supplied as a high-purity solid intended for laboratory research applications.
- High purity (99.92%).
- Molecular formula C17H21ClN4O3 and molecular weight 364.83 g/mol.
- Soluble in DMSO at ≥100 mg/mL.
- Solid, white to off-white, easy to weigh and handle.
- Store sealed at 4°C; solutions stable at -80°C (up to 6 months) or -20°C (up to 1 month).
- Useful as a CRBN ligand for PROTAC and targeted protein degradation studies.
- Available in small milligram package sizes for research use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.