Learn More
Selleck Chemical LLC Lenalidomide S1029-50mg
SDP
Supplier:  Selleck Chemical LLC S102950MG
Encompass_Preferred
Lenalidomide is a TNF- secretion inhibitor with IC50 of 13 nM in PBMCs Lenalidomide (CC-5013) is a ligand of ubiquitin E3 ligase cereblon (CRBN) and it causes selective ubiquitination and degradation of two lymphoid transcription factors IKZF1 and IKZF3 by the CRBN-CRL4 ubiquitin ligase Lenalidomide promotes cleaved caspase-3 expression and inhibit VEGF expression and induces apoptosis
Catalog No. 50-311-1315