Learn More
Medchemexpress LLC Lenalidomide-C4-NH2 25mg | 2435715-90-3 | 351.83 g/mol | C17H22ClN3O3 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY115446A25MG
Encompass_Preferred
Lenalidomide-C4-NH2 hydrochloride is a lenalidomide-derived cereblon ligand supplied as the hydrochloride salt for use as an E3 ligase recruiter in PROTAC synthesis and chemical biology research. It is provided as a solid suitable for conjugation to linkers and target ligands.
  • Hydrochloride salt, solid form suitable for handling and storage.
  • Functions as a cereblon (CRBN) ligand for PROTAC design and E3 ligase recruitment.
  • High purity (HPLC 95.8%) consistent with research-grade material.
  • Available in small research pack sizes, including 25 MG.
  • Molecular formula C17H22ClN3O3; molecular weight ~351.83 g/mol.
Catalog No. 50-002-27070