Learn More
Medchemexpress LLC Lenalidomide-C5-acid | 2338824-30-7 | 96.36% | 373.40 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY16024150MG
Encompass_Preferred
Lenalidomide-C5-acid is a Lenalidomide-based E3 ligase ligand-linker conjugate that recruits the CRBN protein. It can be used to synthesize PROTACs.
  • Recruits the CRBN protein
  • Can be used to synthesize PROTACs
Catalog No. 50-003-94164