Learn More
Medchemexpress LLC Lenalidomide-F | 2359705-88-5 | MFCD31556181 | 99.4% | 262.24 g/mol | C13H11FN2O3 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYW039233
Encompass_Preferred
Lenalidomide-F is a research-grade cereblon E3 ligase ligand commonly used as the recruiting moiety in PROTAC molecules. It facilitates cereblon-mediated, proteasome-dependent degradation of target proteins and has been used in degraders reporting subnanomolar potency (DC50 ≈ 0.64 nM). It is supplied as a solid for biochemical and cellular research applications.
  • E3 ligase ligand that recruits cereblon.
  • Enables proteasome-dependent degradation when incorporated into PROTACs.
  • Demonstrated FLT3 degradation with DC50 ≈ 0.64 nM.
  • Reported high purity suitable for biochemical and cellular assays.
  • Available in multiple pack sizes for research flexibility.
Catalog No. 50-003-03430