Learn More
Medchemexpress LLC HY-131318 100mg Medchemexpress, Lenalidomide-I CAS:2207541-30-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY131318100MG
Encompass_Preferred
Medchemexpress, HY-131318 100mg Lenalidomide-I CAS:2207541-30-6 Lenalidomide-I (Compound 72), an analog of cereblon (CRBN) ligand Lenalidomide for E3 ubiquitin ligase, is used in the recruitment of CRBN protein. Lenalidomide-I can be connected to the ligand for protein by a linker to form PROTACs, such as the PROTAC BET degrader QCA570 (HY-112609). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1271