Learn More
Medchemexpress LLC Lenalidomide-PEG1-azide | 2185795-67-7 | MFCD34469882 | 95.8% | 386.36 g·mol⁻¹ | C17H18N6O5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1331395MG
Encompass_Preferred
Lenalidomide-PEG1-azide is an E3 ligase ligand-linker conjugate that combines a lenalidomide-derived cereblon-binding moiety with a short PEG1 linker terminating in an azide. It is intended as a building block for PROTAC construction and for bioorthogonal conjugation using copper-catalyzed or strain-promoted azide-alkyne cycloaddition.
  • Terminal azide enables CuAAC and SPAAC click chemistry.
  • Lenalidomide-derived cereblon ligand for E3 ligase recruitment.
  • Short PEG1 linker provides minimal steric bulk.
  • High chemical purity suitable for research use.
  • Molecular weight 386.36 g·mol⁻¹; CAS 2185795-67-7.
  • Available in small-scale quantities for synthesis and screening.
Catalog No. 50-002-62501