Learn More
Medchemexpress LLC Lenalidomide-propargyl-C2-NH2 (hydrochloride) | 2489242-23-9 | 98.0% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY130683
Encompass_Preferred
Lenalidomide-propargyl-C2-NH2 hydrochloride incorporates a cereblon (CRBN) ligand for the E3 ubiquitin ligase and a linker. It can be utilized in the design of PROTAC MD-224. As a click chemistry reagent, Lenalidomide-propargyl-C2-NH2 (hydrochloride) contains an Alkyne group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that possess Azide groups.
  • Incorporates a cereblon (CRBN) ligand for the E3 ubiquitin ligase and a linker.
  • Can be used to design the PROTAC MD-224.
  • Contains an Alkyne group for click chemistry.
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that possess Azide groups.
Catalog No. 50-003-41419