Learn More
Medchemexpress LLC Lenalidomide (Standard) | 191732-72-6 | 99.98% | C13H13N3O3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYA0003R100MG
Encompass_Preferred
Lenalidomide (Standard) serves as the analytical standard for Lenalidomide and is designated for research and analytical uses. It is a derivative of Thalidomide that functions as a molecular glue and an orally active immunomodulator. Lenalidomide (CC-5013) acts as a ligand for ubiquitin E3 ligase cereblon (CRBN), leading to the selective ubiquitination and degradation of IKZF1 and IKZF3, two lymphoid transcription factors, by the CRBN-CRL4 ubiquitin ligase. This compound specifically hinders the growth of mature B-cell lymphomas, including multiple myeloma, and stimulates IL-2 release from T cells.
  • Analytical standard: This compound is an analytical standard, used as a reference standard for assays.
  • Research applications: Commonly used in qualitative, quantitative and methodological research experiments.
  • Compatibility: Suitable for use in HPLC, GC, and MS applications.
Catalog No. 50-004-33133