Learn More
Medchemexpress LLC Leniolisib-d5 | 1354690-24-6 | 98.8% | 455.49 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY17635S5MG
Encompass_Preferred
Leniolisib-d5 is a deuterated labeled Leniolisib. Leniolisib (CDZ173) is a potent and selective PI3Kδ inhibitor with potential for treating immunodeficiency disorders. Stable heavy isotopes, such as deuterium, are incorporated into drug molecules as tracers for quantitation during drug development and can influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterated labeled compound, useful as a tracer and internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Potent and selective PI3Kδ inhibitor.
  • Potential for immunodeficiency disorders treatment.
Catalog No. 50-003-87423