Learn More
Medchemexpress LLC Leniolisib (phosphate) | 1354691-97-6 | 99.1% | 548.45 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY17635A100MG
Encompass_Preferred
Leniolisib phosphate is a potent and selective PI3Kδ inhibitor that has the potential for immunodeficiency disorders treatment.
  • Potent and selective PI3Kδ inhibitor.
  • Potential for immunodeficiency disorders treatment.
Catalog No. 50-003-87422