Learn More
Medchemexpress LLC Leniolisib (phosphate) | 1354691-97-6 | 99.10% | 548.45 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY17635A
Encompass_Preferred
Leniolisib (CDZ173) phosphate is a potent and selective PI3Kδ inhibitor. It has potential for treating immunodeficiency disorders.
  • Potent and selective PI3Kδ inhibitor.
  • Potential for immunodeficiency disorders treatment.
  • Suppresses increased pathway activity (measured as phosphorylation of AKT or S6) caused by APDS mutant p110δ in a concentration-dependent manner.
Catalog No. 50-003-84745