Learn More
Takara Bio Lenti-X™ Concentrator, 500 ml
Supplier: Takara Bio 631232
The Lenti-X Concentrator is a complete reagent for the concentration of infectious lentiviral vector particles. This reagent provides a scalable alternative to ultracentrifugation for viral particle concentration. Vector supernatants can be concentrated 10–100 fold, depending upon the volumes used. 500ml