Learn More
Sigma Aldrich Fine Chemicals Biosciences Lentil Lectin SepharoseR 4
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GE17044401
Encompass
Lentil Lectin Sepharose(R) media is used for purification of a number of glycoproteins and other carbohydrate containing molecules. The lentil lectin ligand has more equal affinity for alpha-D-mannose and alpha-D-glucose compared to Con A.Lentil lectin is a metalloprotein containing Ca2 and Mn2 which binds reversibly to polysaccharides and glycoconjugates containing glucose or mannose type sugars. Lentil Lectin Sepharose(R) 4B is a generally applicable group specific adsorbent and is routinely used in the preparation and purification of a number of glycoproteins and carbohydrate containing molecules.
Catalog No. 50-290-8407