Learn More
Apexbio Technology LLC Lentinan 37339-90-5 25mg
SDP
Supplier:  Apexbio Technology LLC C650125
Encompass_Preferred
Lentinan (CAS 37339-90-5) is a polysaccharide compound derived from the fruiting bodies of Lentinus edodes (shiitake mushroom) Lentinan exerts its biological activity primarily through immunomodulation involving the activation of macrophages and natural killer (NK) cells as well as modulation of cytokine secretion Additionally lentinan possesses antioxidative activity which may contribute to protection against oxidative stress-induced cellular damage Based on these pharmacological properties lentinan holds research potential in tumor biology immunotherapy and immune-related disorders
Catalog No. 50-000-89808