Learn More
Takara Bio Lenti-X™ Concentrator - 100ml
SDP
Supplier:  Takara Bio 631231
Encompass
Effortlessly concentrate lentiviral supernatants 100 fold in 1 hour, no ultracentrifugation required. Simply mix and spin. Hassle-free and easily scaled up for large volumes.
Catalog No. NC9833735