Learn More
Apexbio Technology LLC Lenvatinib (E7080), 417716-92-8, MFCD16038644, 10 mM in 1 mL of DMSO
SDP
Supplier:  Apexbio Technology LLC A21745.1
Encompass_Preferred
MF: C21H19ClN4O4, MW: 426.85, Solubility: 21.35mg/mL in DMSO. Lenvatinib (E7080) is an inhibitor of multiple RTK with IC50 values of 22nM, 4nM, 5.2nM, 39nM and 35nM on VEGFR-1, VEGFR-2, VEGFR-3, PDGFRbeta and RET, respectively.
Catalog No. 50-115-0442