Learn More
Medchemexpress LLC Lep(116-130)(mouse) | 258276-95-8 | 99.2% | 1560.73 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP102710MG
Encompass_Preferred
LEP(116-130)(mouse) is a synthetic leptin peptide fragment intended for research use only. In vivo studies have shown that it can lead to a reduced rate of body weight gain, despite increased food intake, and significantly lowers blood glucose levels.
  • Synthetic leptin peptide fragment
  • Unable to inhibit AP-OB binding in vitro
  • Reduces body weight gain
  • Increases food intake
  • Significantly reduces blood glucose levels
  • No effect on thermoregulation in C57BLKS/J-m mice
Catalog No. 50-004-25089