Learn More
Medchemexpress LLC Lepidiline A | 596093-98-0 | 99.5% | 1 ML
SDP
Supplier:  Medchemexpress LLC HY12958310MM/1ML
Encompass_Preferred
Lepidiline A demonstrates cytotoxic activity against HL-60 cells with an IC50 value of 32.3 μM. It targets HSD17B1 activity, enhancing the bioconversion efficiency of sex hormones, including the conversion of estrogen into 17β-estradiol and 4-androsten-3,7-dione into testosterone. This action improves the reproductive capacity of fruit flies. The product is for research use only.
  • Exhibits cytotoxic activity against HL-60 cells with an IC50 value of 32.3 μM
  • Targets HSD17B1 activity
  • Increases the bioconversion efficiency of sex hormones
  • Improves the reproductive capacity of fruit flies
Catalog No. 50-003-42353