Learn More
Medchemexpress LLC Lepidiline A | 596093-98-0 | >99.5% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY129583100MG
Encompass_Preferred
Lepidiline A demonstrates cytotoxic activity against HL-60 cells with an IC50 value of 32.3 μM. It targets HSD17B1 activity, enhancing the bioconversion efficiency of sex hormones, including the conversion of estrogen to 17β-estradiol and 4-androsten-3,7-dione to testosterone. This action contributes to improved reproductive capacity in fruit flies.
  • Targets HSD17B1 activity
  • Enhances sex hormone bioconversion efficiency
  • Improves reproductive capacity in fruit flies
Catalog No. 50-003-38424