Learn More
Medchemexpress LLC Lepidiline A | 596093-98-0 | 99.5% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY12958350MG
Encompass_Preferred
Lepidiline A is a natural product, appearing as a white to off-white solid. It exhibits cytotoxic activity against HL-60 cells and targets 17β-HSD activity, enhancing sex hormone bioconversion. This product is intended for research use only, not for medical applications.
  • Exhibits cytotoxic activity against HL-60 cells with an IC50 value of 32.3 μM.
  • Targets HSD17B1 activity.
  • Increases the bioconversion efficiency of sex hormones.
  • Improves the reproductive capacity of fruit flies.
  • Supplied as a solid with a white to off-white appearance.
  • For research use only.
Catalog No. 50-003-38430