Learn More
Medchemexpress LLC Lepidiline A | 596093-98-0 | 99.54% | 10 MG

Supplier: Medchemexpress LLC HY12958310MG
Lepidiline A exhibits cytotoxic activity against HL-60 cells with an IC50 value of 32.3 μM. It targets HSD17B1 activity, increasing the bioconversion efficiency of sex hormones (including the conversion of estrogen into 17β-estradiol and 4-androsten-3,7-dione into testosterone), which in turn improves the reproductive capacity of fruit flies.
- Targets HSD17B1 activity
- Enhances sex hormone bioconversion
- Exhibits cytotoxic activity against HL-60 cells
- Stable as powder at -20°C for 3 years
- Stable in solvent at -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.