Learn More
Medchemexpress LLC Leptin Human His 2ug
SDP
Supplier:  Medchemexpress LLC HYP7232A2UG
Encompass_Preferred
Leptin is a cytokine-like sugar secretory protein secreted by adipocytes that helps regulate energy balance by inhibiting food intake and increasing thermogenesis Leptin can activate the JAK2-STAT3 and PI3K-AKT/mTOR pathways to regulate energy metabolism prolong the QT interval via MAPK in the myocardium and play a protective role in the gastric mucosa by inducing NO/CGRP release via the vagus nerve Circulating CRP protein binds to leptin and blocks signal transduction leading to obesity resistance Leptin Protein Human (His) is a recombinant human Leptin protein expressed by E coli with N-6 His tag
Catalog No. 50-001-82800