Learn More
Enzo Life Sciences Leptin (mouse), ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI900019A
Encompass_Preferred
The Leptin (mouse), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 3 hours. Absorbance is read at 450 nm. The broad dynamic range is suitable for many normal and pathological samples with sensitive quantification of < 26 pg/mL mouse leptin. Alternative Name: OB gene product. Sensitivity: 25.4 pg/ml (range 50 - 3,200 pg/ml). Assay Time: 3 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Mouse. Contents: Microtiter plate, Standard, Antibody, Conjugate, Assay buffer 28, TMB Substrate, Stop solution 2, Wash buffer concentrate
Catalog No. 50-658-105