Learn More
Medchemexpress LLC Leptomerine | 22048-97-1 | MFCD28336213 | 99.9% | 201.26 g/mol | C13H15NO | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN420610MG
Encompass_Preferred
Leptomerine is a naturally occurring quinolinone alkaloid that inhibits acetylcholinesterase and is supplied as a high-purity research compound for in vitro and neuropharmacology studies.
  • High purity (99.9%) suitable for research use.
  • Molecular formula C13H15NO; molecular weight 201.26 g/mol.
  • Solid form, convenient for storage and weighing.
  • Reported acetylcholinesterase inhibitory activity; useful in neurodegenerative disease research and biochemical assays.
  • Available in milligram-scale quantities for small-scale experiments.
  • COA and SDS documentation available from the supplier.
Catalog No. 50-002-89146