Learn More
ATCC Leptospira; interrogans; serovar Copenhageni
Supplier:  ATCC BAA1198
Encompass
Leptospira interrogans serovar Copenhageni strain Fiocruz L1-130 is a whole-genome sequenced bacterium that was isolated from a patient with leptospirosis.
Catalog No. 50-238-1909
 Restricted Item