Learn More
Medchemexpress LLC LEQ506 | 1204975-42-7 | 98.9% | 432.56 | 100 MG

Supplier: Medchemexpress LLC HY18636100MG
LEQ506 is a second-generation inhibitor of smoothened (Smo) with IC50 values of 2 nM in human and 4 nM in mouse. It inhibits Hedgehog (Hh) signaling in a human cell line (HEPM) by decreasing Gli mRNA, with an IC50 approximately 6-fold lower than that of Compound 2. At 1%, LEQ506 achieves 80% to 90% inhibition for Gli1 and 60% to 70% for Ptch1.
- Inhibits smoothened (Smo)
- Has IC50 values of 2 nM in human and 4 nM in mouse
- Inhibits Hedgehog (Hh) signaling
- Decreases Gli1 mRNA by 70% to 80%
- Preferentially inhibits Gli1 over Ptch1 mRNA
- Is an efficacious compound
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.