Learn More
Medchemexpress LLC Lercanidipine | 100427-26-7 | 99.9% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB061210MM/1ML
Encompass_Preferred
Lercanidipine is a third-generation, lipophilic, brain-penetrant, vascular-selective, and orally active dihydropyridine-calcium channel blocker. It exhibits long-lasting antihypertensive action and reno- and neuro-protective effects. This product is suitable for use in cardiovascular and neurological research.
  • Has a pIC50 of 7.74
  • Offers reno- and neuro-protective effects
  • Exhibits anti-oxidant, anti-inflammatory, and anti-apoptotic properties
  • Used in cardiovascular and neurological research
Catalog No. 50-004-08384