Learn More
Medchemexpress LLC Lercanidipine | 100427-26-7 | 99.9% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB0612
Encompass_Preferred
Lercanidipine is a third-generation, lipophilic, brain-penetrant, vascular-selective, and orally active dihydropyridine-calcium channel blocker. It exhibits long-lasting antihypertensive action and possesses reno- and neuro-protective effects, making it suitable for cardiovascular and neurological research.
  • Vascular-selective calcium channel blocker
  • Orally active with long-lasting antihypertensive action
  • Possesses reno- and neuro-protective effects
  • Demonstrates anti-oxidant, anti-inflammatory, and anti-apoptotic properties
Catalog No. 50-004-50488