Learn More
Medchemexpress LLC Lercanidipine | 100427-26-7 | 99.9% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB061250MG
Encompass_Preferred
Lercanidipine is a third-generation, lipophilic, brain-penetrant, vascular-selective, and orally active dihydropyridine-calcium channel blocker with a pIC50 of 7.74. It has a long-lasting antihypertensive action and also exhibits reno- and neuro-protective effects. Additionally, Lercanidipine demonstrates anti-oxidant, anti-inflammatory, and anti-apoptotic properties. It is suitable for cardiovascular and neurological research.
  • Third-generation dihydropyridine-calcium channel blocker
  • Lipophilic and brain-penetrant
  • Vascular-selective and orally active
  • Long-lasting antihypertensive action
  • Reno- and neuro-protective effect
  • Anti-oxidant, anti-inflammatory, and anti-apoptotic properties
  • Can be used in cardiovascular and neurological research
Catalog No. 50-004-05882