Learn More
Apexbio Technology LLC Lercanidipine 100mg
SDP
Supplier:  Apexbio Technology LLC B3483100
Encompass_Preferred
Lercanidipine (CAS 100427-26-7) is a synthetic dihydropyridine derivative that functions as a selective inhibitor of voltage-gated L-type calcium channels By blocking calcium influx into vascular smooth muscle cells it induces vasodilation and reduces peripheral vascular resistance Lercanidipine is widely utilized in research investigating calcium-mediated signal transduction vascular reactivity and hypertension-related pathophysiology Its well-characterized pharmacological properties make it a valuable tool for elucidating mechanisms underlying cardiovascular regulation and for evaluating novel interventions targeting calcium channel dynamics
Catalog No. 50-000-96338