Learn More
TARGETMOL CHEMICALS INC LERCANIDIPINE 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T251910MG
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Lercanidipine (Masnidipine) is a calcium channel blocker of the dihydropyridine class. purity: 99%
Catalog No. 50-257-7099