Learn More
TARGETMOL CHEMICALS INC Lerociclib dihydrochloride
SDP
Supplier:  TARGETMOL CHEMICALS INC T11345L10MG
Encompass_Preferred
Also available in 1 mg, 2 mg, 5 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Lerociclib dihydrochloride (G1T38 dihydrochloride) is a potent and selective CDK4/CDK6 inhibitor, with IC50 values of 2 nM for CDK6/CyclinD3 and 1 nM for CDK4/CyclinD1. Purity 97.4%
Catalog No. 50-288-0661